Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SURF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567356 Supplier Invitrogen™ Supplier No. PA567356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEKRQRVKGNL TPLTGRNYRQ LLERLQARQS RLDELRGQDE GKAQELEAKM KWTNLLYKAE GVKIRDDERL L Highest antigen sequence identity to the following orthologs - mouse 92%, rat 89%.

This gene is located in the surfeit gene cluster, a group of very tightly linked genes that do not share sequence similarity. The gene demonstrates features of a housekeeping gene, being ubiquitously expressed, and the encoded protein has been localized to the nucleolus. The protein includes motifs found in both the mouse and fish orthologs, which suggests a putative function as a nucleolar-matrix protein with nucleic acid-binding properties, based on characteristics determined in mouse.This gene is located in the surfeit gene cluster, a group of very tightly linked genes that do not share sequence similarity. The gene demonstrates features of a housekeeping gene, being ubiquitously expressed, and the encoded protein has been localized to the nucleolus. The protein includes motifs found in both the mouse and fish orthologs, which suggests a putative function as a nucleolar-matrix protein with nucleic acid-binding properties, based on characteristics determined in mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SURF6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SURF6
Gene Accession No. O75683
Gene Alias D2Wsu129e; RRP14; SURF6; Surf-6; surfeit 6; surfeit gene 6; surfeit locus protein 6
Gene Symbols SURF6
Host Species Rabbit
Immunogen Recombinant Human SURF6. Recombinant protein control fragment (Product #RP-105590).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6838
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.