Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surface Lipoprotein p27 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990904
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0904 100 μg
50-199-0905 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0904 Supplier Rockland Immunochemicals Supplier No. 200401C30
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. It is directed against, and shows specific reactivity for, Borrelia burgdorferi p27 protein. Reactivity with p27 protein from other sources has not been determined.

Surface Lipoprotein p27 of Borrelia burgdorferi is a surface-exposed lipoprotein that has been shown (by Western blot and Northern blot) to be expressed in the European B. burgdorferi strain B29, but not in the American strain B31. Cell envelope proteins of bacterial pathogens play important roles in the host-parasite interactions that occur during infection, including cell adherence, cell invasion, and immune cell activation or evasion. p27 is a basic protein of 248 amino acids with a typical prokaryotic leader sequence of 17 amino acid residues at the N-terminus of the proposed translation product. The p27 gene is located on a linear plasmid of a size of approximately 55 kb. Borrelia spirochetes are unique among diderm bacteria in their abundance of surface-displayed lipoproteins, some of which play important roles in the pathogenesis of Lyme disease and relapsing fever. There is evidence that Borrelia lipoproteins are specifically targeted to the bacterial surface, but that they can be retained in the periplasm by sequence-specific signals.

Specifications

Antigen Surface Lipoprotein p27
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen MBP-fusion protein corresponding to Borrelia burgdorferi Surface Lipoprotein p27 protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.