Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Surfactant Protein D, Rabbit, Alexa Fluor 488, Polyclonal,

Catalog No. ABS574AF488 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS574AF488 100 μL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS574AF488 Supplier MilliporeSigma™ Supplier No. ABS574AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pulmonary surfactant-associated protein D (UniProt P35247; also known as Collectin-7, Lung surfactant protein D, PSP-D, SP-D) is encoded by the SFTPD (also known as COLEC7, PSPD, SFTP4) gene (Gene ID 6441) in human. Surfactant, produced by alveolar type II cells and airway Clara cells, resides at the air-liquid interface of the lungs and is most commonly known for its role in reducing surface tension. Surfactant is a lipoprotein complex composed of approximately 10% proteins, including surfactant protein A (SP-A), SP-B, SP-C, and SP-D. SP-A and SP-D belong to the collectin family of proteins and can modulate innate immunity. SP-D is known to enhance pulmonary clearance of pathogens, such as Pseudomonous aerginosa, Klebsiella pneumonia, respiratory syncytial virus (RSV) and Influenza virus. SP-D is also shown to modify allergic responsee in the lungs by binding common allergens, including house dust mite and pollen particles. SP-A-deficient and SP-D-deficient mice are susceptible to bleomycin-induced lung injury and display increased cellular inflammation, more severe lung fibrosis, and reduced survival. In addition to lung, SP-D is also reported to be detected in extrapulmonary tissues, such as kidney and pancreas. Serum levels of either SP-A or SP-D in human are correlated with mortality in patients with pulmonary fibrosis. Human SP-D is produced with a signal peptide sequence (a.a. 1-20), which is removed posttranslationally to yield the mature protein (a.a. 21-375) that contains a collagen-like domain (a.a. 46-222), a coiled coil region (a.a. 223-252), and a C-type lectin domain (a.a. 260-375).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Surfactant Protein D
Applications Immunofluorescence
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 488
Formulation Purified rabbit polyclonal antibody conjugate in PBS with 1.5% BSA and 0.05% Sodium Azide.
Gene SFTPD, COLEC7, PSPD, SFTP4
Gene Accession No. P35247
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence within the C-type lectin domain of human surfactant protein D.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_003010
Test Specificity This Alexa Fluor™ 488-conjugated polyclonal antibody targets a sequence within the C-type lectin domain of human surfactant protein D.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.