Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surfactant protein D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501727496 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7496 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7496 Supplier Proteintech Group Inc Supplier No. 118391AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Surfactant protein D (SP-D) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular ′head′ regions linked by triple-helical, collagen-like, strands. This group also includes SP-A and the serum proteins man-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages.

Specifications

Antigen Surfactant protein D
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SFTPD
Gene Accession No. P35247, P35248, P50404
Gene Alias COLEC7, Collectin 7, Lung surfactant protein D, PSP D, PSPD, SFTP4, SFTPD, SP D, surfactant protein D
Gene Symbols SFTPD
Host Species Rabbit
Immunogen Surfactant protein D Fusion Protein Ag2461
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20390, 25350, 6441
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.