Learn More
Surfeit 2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304611AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene shares a bidirectional promoter with surfeit 1 (SURF1), which is located on the opposite strand. It encodes a conserved protein that is expressed in a variety of tissues.Specifications
Surfeit 2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q15527 | |
SURF2 | |
Between 175 and 225 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SURF2 | |
SURF-2, surfeit locus protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6835 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.