Learn More
Surgical Design No. 12 Carbon Scalpel Blade
Catalog No. 22079692 Shop All Surgical Design Products
Click to view available options
Size:
# 12
This item is not returnable. View return policy
Catalog No. 22-079-692
Supplier: Surgical Design BLADE#12
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Small, pointed, crescent shaped blade fits handles 3, 3L, 3 Graduated, 5B, 7, 9, B3 and B3L
  • Sharpened along inside edge of curve
  • Used for surgical incision of an artery
  • Used for calculus removal by incision of ureter
  • Used for surgical incision of renal pelvis of kidney for removal of kidney stone
Specifications
Size # 12
For Use With (Equipment) Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.