Learn More
Surgical Design No. 20 Carbon Scalpel Blade
Catalog No. 22079695 Shop All Surgical Design Products
This item is not returnable. View return policy
Catalog No. 22-079-695
Supplier: Surgical Design BLADE#20
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Curved cutting edge blade fits handles 4, 4L, 4 Graduated and 6B
  • Large version of No.10 blade
  • Flat, unsharpened back edge
  • Used for orthopaedic and general surgical procedures
Specifications
For Use With (Equipment) Handles # 4, # 4L, # 4 Graduated and # 6B
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.