Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Surgical Knife with Comfort Grip, Scalpel Handle

Catalog No. p-200063787 Shop All Surgical Design Products
Click to view available options
Size:
# 3
# 4
Color:
Blue
Green
Orange
Pink
Purple
Red
This item is not returnable. View return policy
This item is not returnable. View return policy

Surgical Design™ Surgical Knife Scalpel Handle used for sharp and precise dissection. The knife handle comes with enhanced PVC comfort and secure grip that increases precision during use.

  • Surgical Design™ Premium products exceed performance and quality expectations
  • Handles ergonomically designed for precision work
  • Handles offer color identification
  • Comfortable, reusable
  • Blades slide on smoothly and securely
  • Superb quality and workmanship
  • Premium surgical grade stainless steel
  • Corrosion protected by passivation and ultrasonic cleaning
  • Sterilizable by EtOH/Gamma Radiation
  • Fully autoclavable

Specifications

Product Type Scalpel Handle (High Precision)
For Use With (Application) 20 Series Scalpel Blades
Handle Type Comfort Grip
Size # 4
Disposable Reusable
Sterility Non-sterile
Certifications/Compliance ISO, GMP
Autoclavable Autoclavable
Color Red
Graduated No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.