Learn More
Terumo Medical Corporation Surshield-PUR Safety I.V. Catheters 22G
Catalog No. p-200136074 Shop All Terumo Medical Corporation Products
Click to view available options
Length (English):
1 1/4 in.
1 in.
This item is not returnable. View return policy
This item is not returnable. View return policy
Safety I.V. Catheters.
  • Passive safety to reduce risk of needlestick injury, without adding extra steps to your procedures
  • SurFlash™ enhanced flashback feature provides clear visual confirmation of catheter placement, for increased confidence and first-stick success
  • Easy to use
  • Automatic activation - no extra steps needed
  • A compact design that minimizes the waste volume
Specifications
Material Polyurethane, Polypropylene, Stainless Steel (Needle)
Needle Gauge 22 G
Latex Free Yes
Length (English) 1 in.
Product Type IV Catheter
Disposable Single-use
For Use With (Equipment) IV Lines
Connection Type Catheter
Graduated No
Sterility Sterile
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.