Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terumo™ Surshield™ Safety Winged Infusion Sets

Catalog No. p-322968
Click to view available options
Needle Gauge:
19 G
21 G
23 G
25 G
Length (English) Tube:
3.5 in.
12 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

One-handed activation for IV infusion therapy

  • Nonlatex plastic and tubing
  • Allergy-free usage
  • Flexible wings
  • Polyvinyl tubing
  • Siliconized needle

Specifications

Product Type Safety Winged Infusion Set
Material PVC Tubing, Siliconized Needle
Needle Gauge 25 G
Sterility Sterile
For Use With (Application) IV infusion therapy
Length (English) Needle 0.75 in.
Length (English) Tube 3.5 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.