Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Survivin Antibody (32.1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 11 publications
$207.00 - $499.50
Specifications
Antigen | Survivin |
---|---|
Clone | 32.1 |
Concentration | 1.9 mg/mL |
Dilution | Western Blot 1:1000, Flow Cytometry, ELISA reported in scientific literature (PMID 11861764), Immunohistochemistry 20 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation reported in scientific literature (PMID 11861764), Immunohistochemistry-Paraffin 20 ug/ml |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB500237SS
![]() |
Novus Biologicals
NB500237SS |
0.025 mL |
Each for $207.00
|
|
|||||
NB500237
![]() |
Novus Biologicals
NB500237 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Survivin Monoclonal specifically detects Survivin in Human, Hamster samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Survivin | |
1.9 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells | |
O15392 | |
332 | |
This Survivin Antibody (32.1) was developed against full length recombinant human Survivin [UniProt# O15392] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
16 kDa |
32.1 | |
Western Blot 1:1000, Flow Cytometry, ELISA reported in scientific literature (PMID 11861764), Immunohistochemistry 20 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation reported in scientific literature (PMID 11861764), Immunohistochemistry-Paraffin 20 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Hamster | |
API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
BIRC5 | |
IgG1 κ | |
Protein G purified | |
Survivin Antibody (32.1) [NB500-237] is specific for the nuclear form of survivin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title