Learn More
Survivin Antibody (60.11) - BSA Free, Novus Biologicals™
SDP
Mouse Monoclonal Antibody has been used in 42 publications
Supplier:  Novus Biologicals NB500238
 View more versions of this product
Catalog No. NB500238
Only null left
Add to Cart
Description
Description
Survivin Monoclonal specifically detects Survivin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Flow (Intracellular), Knockdown Validated.
Specifications
Specifications
Survivin
Monoclonal
0.9 mg/mL
Western Blot 1:1000, Flow Cytometry 1 ug/mL. Use reported in scientific literature (PMID 25050620), ELISA reported in scientific literature (PMID 11861764), Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen, Proximity Ligation Assay reported in scientific literature (PMID 28077791), Flow (Intracellular), KnockDown Validated
API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
Mouse
16 kDa
0.1 mL
Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
332
Human, Mouse, Rat
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
60.11
Unconjugated
O15392
BIRC5
This Survivin Antibody (60.11) [Alexa Fluor 350] was developed against full length recombinant human Survivin [UniProt# O15392]
Protein G purified
RUO
Primary
Survivin Antibody (60.11) [NB500-238] is specific for the cytoplasmic form of Survivin.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG2a κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only