Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Survivin Antibody (60.11) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500238SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500238SS 0.025 mL
NB500238 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500238SS Supplier Novus Biologicals Supplier No. NB500238SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 42 publications

Survivin Monoclonal specifically detects Survivin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Flow (Intracellular), Knockdown Validated.

Specifications

Antigen Survivin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 60.11
Concentration 0.9 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1 ug/mL. Use reported in scientific literature (PMID 25050620), ELISA reported in scientific literature (PMID 11861764), Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen, Proximity Ligation Assay reported in scientific literature (PMID 28077791), Flow (Intracellular), KnockDown Validated
Gene Accession No. O15392
Gene Alias API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
Gene Symbols BIRC5
Host Species Mouse
Immunogen This Survivin Antibody (60.11) [Alexa Fluor 350] was developed against full length recombinant human Survivin [UniProt# O15392]
Molecular Weight of Antigen 16 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 332
Test Specificity Survivin Antibody (60.11) [NB500-238] is specific for the cytoplasmic form of Survivin.
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.