Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Survivin Antibody (60.11) - Unpurified, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 22 publications

$206.50 - $426.00

Specifications

Antigen Survivin
Clone 60.11
Dilution Western Blot reported in scientific literature (PMID 18332262), Flow Cytometry 1:200, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 18332262), Immunohistochemistry-Frozen reported in scientific literature (PMID 22841823), Proximity Ligation Assay reported in scientific literature (PMID 28077791), KnockDown Validated
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA, KnockDown
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB500205SS
SDP
View Documents
Novus Biologicals
NB500205SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB500205
SDP
View Documents
Novus Biologicals
NB500205
0.1 mL
Each for $426.00
Only null left
Add to Cart
 
Description

Description

Survivin Monoclonal specifically detects Survivin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Knockdown Validated.
Specifications

Specifications

Survivin
Western Blot reported in scientific literature (PMID 18332262), Flow Cytometry 1:200, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 18332262), Immunohistochemistry-Frozen reported in scientific literature (PMID 22841823), Proximity Ligation Assay reported in scientific literature (PMID 28077791), KnockDown Validated
Monoclonal
Ascites
RUO
Human, Mouse, Rat
O15392
332
This Survivin Antibody (60.11) was developed against full length recombinant human Survivin [UniProt# O15392].
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
16 kDa
60.11
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA, KnockDown
Unconjugated
Mouse
Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
Ascites with PBS with 0.05% Sodium Azide
API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
BIRC5
IgG2a κ
Unpurified
Survivin Antibody (60.11) [NB500-205] is specific for the cytoplasmic form of survivin.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.