Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Survivin Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB500201H0.1ML
Description
Survivin Polyclonal specifically detects Survivin in Human, Mouse, Rat, Canine, Feline, Guinea Pig, Hamster samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.Specifications
Survivin | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
API4 survivin variant 3 alpha, Apoptosis inhibitor 4 baculoviral IAP repeat-containing protein 5, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, BIRC5, EPR-1, IAP4 | |
Rabbit | |
Affinity Purified | |
332 | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
HRP | |
PBS | |
BIRC5 | |
Full length recombinant human Survivin [UniProt# O15392] | |
0.1 mL | |
Human, Mouse, Rat, Canine, Feline, Guinea Pig, Hamster | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction