Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HS2ST1 Polyclonal Antibody, Cyanine3, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS17389RCY3

This item is not returnable.
View return policy
Description
HS2ST1 Polyclonal antibody specifically detects HS2ST1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)Specifications
HS2ST1 | |
Polyclonal | |
Cyanine3 | |
HS2ST1 | |
2OST; 2-OST; 2-O-sulfotransferase; AW214369; dJ604K5.2; heparan sulfate 2-O-sulfotransferase 1; Hs2st; Hs2st1; KIAA0448; mKIAA0448 | |
KLH conjugated synthetic peptide derived from human HS2ST1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
Q7LGA3, Q8R3H7 | |
Rabbit | |
Protein A | |
RUO | |
23908, 9653 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction