Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Survivin Monoclonal Antibody (Mix-mA), Cyanine3, Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM33195MCY3

This item is not returnable.
View return policy
Description
Survivin Monoclonal antibody specifically detects Survivin in Human, Rat samples. It is validated for ImmunocytochemistrySpecifications
Survivin | |
Monoclonal | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
O15392, Q9JHY7 | |
Mouse | |
Protein A | |
RUO | |
332, 64041 | |
-20°C or -80°C if preferred | |
Liquid |
Immunocytochemistry | |
Mix-mA | |
Cyanine3 | |
Birc5 | |
AAC-11; AP14; Api4; apoptosis inhibitor 4; Apoptosis inhibitor survivin; baculoviral IAP repeat containing 5; baculoviral IAP repeat-containing 5; baculoviral IAP repeat-containing 5 (survivin); baculoviral IAP repeat-containing protein 5; BIRC 5; BIRC5; EPR 1; EPR-1; Iap4; inhibitor of apoptosis; survivin; survivin variant 3 alpha; survivin40; SVV; TIAP | |
Recombinant human Survivin | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction