Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Survivin Monoclonal Antibody (STLALYV), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Catalog No. 53917682
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-917-682 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-917-682 Supplier Invitrogen Supplier No. 53917682

Mouse Monoclonal Antibody

Description: This STLALYV monoclonal antibody recognizes human survivin (also known as Birc5). Survivin is a member of the inhibitor of apoptosis (IAP) family that also includes XIAP, c-IAP1, c-IAP2, and NAIP. Survivin functions to inhibit caspase activation, thereby leading to negative regulation of apoptosis. Survivin also plays a critical role in mitosis and cytokinesis where it is expressed only during G2/M and localizes to the mitotic spindle and is found at the cleavage furrow/midplate. Thus, survivin is highly expressed in tumors and in fetal tissue, but is absent in terminally-differentiated cells and non-proliferating cells. Deletion of survivin in developing thymocytes as well as in mature T cells showed that survivin is critical for cell division in lymphoid cells but not for inhibition of survival; the same is true for B lymphocytes. Applications Reported: This STLALYV antibody has been reported for use in microscopy, and immunocytochemistry. Applications Tested: This STLALYV antibody has been tested by immunocytochemistry of fixed and permeabilized cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

Survivin (IAP4) is the smallest member of the Inhibitors of Apoptosis Protein (IAP) gene family. The IAPs are involved in multiple cell functions such as cell signaling, cell division, metabolism, and exhibits differential expression in nearly all human cancers, but not in most normal tissues. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but the Survivin gene encodes proteins with only a single BIR domain. The encoded proteins also lack a C-terminus RING finger domain. Gene expression is high during fetal development and in most tumors yet low in adult tissues. Antisense transcripts are involved in the regulation of survivin’s gene expression. Survivin is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. At the beginning of mitosis, survivin associates with microtubules of the mitotic spindle in a specific and saturable reaction that is regulated by microtubule dynamics. Disruption of survivin-microtubule interactions results in loss of survivin's anti-apoptosis function and increased caspase-3 activity, a mechanism involved in cell death, during mitosis. Survivin may counteract a default induction of apoptosis in G2/M phase. Survivin is also abundantly expressed in brain tissues (astrocytes and some neurons) of adult rats following traumatic brain injury. Survivin has been found co-expressed with NeuN (mature neuronal marker) and PCNA (a cell cycle protein). Survivin might affect regulation of neural cell proliferative responses after brain injury. At least four transcript variants encoding distinct isoforms have been found for this gene, but the full-length natures of only three of them have been determined. The overexpression of survivin in cancer may overcome this apoptotic checkpoint and favor aberrant progression of transformed cells through mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Survivin
Applications Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone STLALYV
Concentration 0.5 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Birc5
Gene Accession No. O15392
Gene Alias AAC-11; AP14; Api4; apoptosis inhibitor 4; Apoptosis inhibitor survivin; baculoviral IAP repeat containing 5; baculoviral IAP repeat-containing 5; baculoviral IAP repeat-containing 5 (survivin); baculoviral IAP repeat-containing protein 5; BIRC 5; BIRC5; EPR 1; EPR-1; Iap4; inhibitor of apoptosis; survivin; survivin variant 3 alpha; survivin40; SVV; TIAP
Gene Symbols Birc5
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 332
Target Species Human
Content And Storage 4° C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.