Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Survivin, p Thr34 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 9 publications
$206.50 - $499.50
Specifications
Antigen | Survivin (p Thr34) |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000. Use reported in scientific literature (PMID 25298395), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23549285) |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB500236SS
![]() |
Novus Biologicals
NB500236SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB500236
![]() |
Novus Biologicals
NB500236 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Survivin Polyclonal specifically detects Survivin in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Survivin (p Thr34) | |
Western Blot 1:1000. Use reported in scientific literature (PMID 25298395), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23549285) | |
Polyclonal | |
Rabbit | |
Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
BIRC5 | |
IgG | |
Affinity Purified | |
Survivin [p Thr34] Antibody [NB500-236] is specific for phosphorylated survivin. |
1 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse | |
O15392 | |
332 | |
This Survivin [p Thr34] Antibody was developed against a synthetic peptide with a phosphorylated Threonine (amino acid 34) corresponding to human Survivin. [UniProt# O15392] | |
Primary | |
Store at 4C. Do not freeze. | |
16 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title