Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUSD2 Antibody (1279C), Allophycocyanin, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB90565APC
Description
SUSD2 Monoclonal specifically detects SUSD2 in Human samples. It is validated for Flow Cytometry.Specifications
SUSD2 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
SUSD2 | |
Partial recombinant human SUSD2 protein (amino acids 1-785) [UniProt Q9UGT4] | |
100 μL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1279C | |
Flow Cytometry | |
BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
Rabbit | |
Protein A or G purified | |
RUO | |
56241 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction