Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUSD2 Antibody (944827), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB90561APCCY7
Description
SUSD2 Monoclonal antibody specifically detects SUSD2 in Human samples. It is validated for Flow CytometrySpecifications
| SUSD2 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 944827 | |
| Flow Cytometry | |
| BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
| Human embryonic kidney cell line HEK293-derived human SUSD2, Met1-Ala785, Accession # Q9UGT4 | |
| 0.1 mL | |
| Cancer | |
| 56241 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction