Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUSD2 Mouse anti-Human, Alexa Fluor™ 647, Clone: W5C5, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566657
Description
SUSD2 Monoclonal antibody specifically detects SUSD2 in Human samples. It is validated for Flow Cytometry.
Specifications
| SUSD2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| SUSD2 | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| W5C5 | |
| Alexa Fluor 647 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction