Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUSD2 Rat anti-Human, Unconjugated, Clone: 19E8, MilliporeSigma™

Catalog No. MABC17142MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC17142MI 25 μL
MABC17141MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC17142MI Supplier MilliporeSigma Supplier No. MABC171425UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SUSD2 Monoclonal antibody specifically detects SUSD2 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUSD2
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 19E8
Conjugate Unconjugated
Gene Accession No. NP_062547.1
Gene Alias Sushi domain-containing protein 2
Gene Symbols SUSD2
Host Species Rat
Immunogen An expression construct corresponding to the extracellular domain of human SUSD2.
Molecular Weight of Antigen 90.21 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 19E8 is a rat monoclonal antibody that detects human Sushi domain-containing protein 2. It targets an epitope within the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.