Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUSD3 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297966APC
Description
SUSD3 Polyclonal antibody specifically detects SUSD3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SUSD3 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
MGC26847, sushi domain containing 3, sushi domain-containing protein 3 | |
Produced in rabbits immunized with E. coli-derived Human SUSD3 fragment. | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
203328 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction