Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SUSD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544351
Description
Peptide sequence: LRLPPQATFQ VLRGNGASVG TVLMFRCPSN HQMVGSGLLT CTWKGSIAEW Sequence homology: Cow: 77%; Human: 100%; Pig: 77%.
SUSD3 is a protein coding gene. May play a role in breast tumorigenesis by promoting estrogen-dependent cell proliferation, cell-cell interactions and migration.
Specifications
| SUSD3 | |
| Polyclonal | |
| Unconjugated | |
| SUSD3 | |
| 1700017I11Rik; 2810440J20Rik; Susd3; sushi domain containing 3; sushi domain-containing protein 3; UNQ9387/PRO34275 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 203328 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96L08 | |
| SUSD3 | |
| Synthetic peptide directed towards the N-terminal of human SUSD3 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction