Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUSD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543029 Supplier Invitrogen™ Supplier No. PA543029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MYHGMNPSNG DGFLEQQQQQ QQPQSPQRLL AVILWFQLAL CFGPAQLTGG Sequence homology: Cow: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 85%; Rat: 93%; Yeast: 92%.

SUSD4 contains 4 Sushi (CCP/SCR) domains. It is a single-pass type I membrane protein. The function of the SUSD4 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUSD4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SUSD4
Gene Accession No. Q5VX71
Gene Alias AI848994; E430021N18Rik; N28096; PRO222; RGD1564043; SUSD4; sushi domain containing 4; sushi domain-containing protein 4; testicular secretory protein Li 55; UNQ196/PRO222; YHGM196
Gene Symbols SUSD4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SUSD4 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55061
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.