Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SUSD5 Monoclonal Antibody (4G3)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524447
Description
Peptide Sequence: TTVCSKGSGE QQIMRAVDVR IESNPVPGGT YSALCIKDEE KPCGDPPSFP HTILQGRTGL EMGDELLYVC APGHIMGHRE TAFTLLCNSC GEWYGLVQAC.
SUSD5 is a protein coding gene and gene ontology (GO) annotation includes integral component of membrane; hyaluronic acid binding; cell adhesion; Notch signaling pathway.
Specifications
SUSD5 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
O60279 | |
SUSD5 | |
SUSD5 (XP_171054, 284 a.a. ~ 383 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4G3 | |
Unconjugated | |
SUSD5 | |
KIAA0527; SUSD5; sushi domain containing 5; sushi domain-containing protein 5 | |
Mouse | |
Protein A | |
RUO | |
26032 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction