Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUV39H1 Monoclonal Antibody (MG44), Active Motif™
SDP

Catalog No. 501992850
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2850 100 μg
50-199-2851 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2850 Supplier Active Motif Supplier No. 39785
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes an evolutionarily-conserved protein containing an N-terminal chromodomain and a C-terminal SET domain. The encoded protein is a histone methyltransferase that trimethylates lysine 9 of histone H3, which results in transcriptional gene silencing. Loss of function of this gene disrupts heterochromatin formation and may cause chromosome instability. Alternative splicing results in multiple transcript variants.

Specifications

Antigen SUV39H1
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MG44
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene SUV39H1
Gene Accession No. O43463, O54864
Gene Alias AI852103; AL022883; DXHXS7466e; H3-K9-HMTase 1; Histone H3-K9 methyltransferase 1; Histone-lysine N-methyltransferase SUV39H1; histone-lysine N-methyltransferase, H3 lysine-9 specific 1; KMT1A; Lysine N-methyltransferase 1A; MG44; mIS6; Position-effect variegation 3-9 homolog; RGD1565028; Su(var)3-9 homolog 1; Suppressor of variegation 3-9 homolog 1; suppressor of variegation 3-9 homolog 1 (Drosophila); SUV39H; Suv39h histone methyltransferase; SUV39H1
Gene Symbols SUV39H1
Host Species Mouse
Immunogen This SUV39H1 antibody was raised against a recombinant protein corresponding to amino acids 1-126 of mouse SUV39H1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20937, 6839
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.