Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUV39H1 Mouse anti-Monkey, Human, Clone: MG44, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $463.74
Specifications
Antigen | SUV39H1 |
---|---|
Clone | MG44 |
Concentration | 0.5 mg/mL |
Applications | ChIP Assay, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
05615I25UL
|
MilliporeSigma
05615I25UL |
25 μL |
Each for $182.31
|
|
|||||
05615I100UL
|
MilliporeSigma
05615I100UL |
100 μL |
Each for $463.74
|
|
|||||
Description
SUV39H1 Monoclonal antibody specifically detects SUV39H1 in Monkey, Human samples. It is validated for ChIP assay (ChIP), Immunoprecipitation, Western Blotting.Specifications
SUV39H1 | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG | |
Protein G purified |
MG44 | |
ChIP Assay, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Monkey, Human | |
MBP-tagged full-length recombinant human Histone-lysine N-methyltransferase SUV39H1. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title