Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUV39H1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986737
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6737 Supplier Bioss Supplier No. BS5708R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes an evolutionarily-conserved protein containing an N-terminal chromodomain and a C-terminal SET domain. The encoded protein is a histone methyltransferase that trimethylates lysine 9 of histone H3, which results in transcriptional gene silencing. Loss of function of this gene disrupts heterochromatin formation and may cause chromosome instability. Alternative splicing results in multiple transcript variants.

Specifications

Antigen SUV39H1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SUV39H1
Gene Accession No. O43463
Gene Alias AI852103; AL022883; DXHXS7466e; H3-K9-HMTase 1; Histone H3-K9 methyltransferase 1; Histone-lysine N-methyltransferase SUV39H1; histone-lysine N-methyltransferase, H3 lysine-9 specific 1; KMT1A; Lysine N-methyltransferase 1A; MG44; mIS6; Position-effect variegation 3-9 homolog; RGD1565028; Su(var)3-9 homolog 1; Suppressor of variegation 3-9 homolog 1; suppressor of variegation 3-9 homolog 1 (Drosophila); SUV39H; Suv39h histone methyltransferase; SUV39H1
Gene Symbols SUV39H1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Histone-lysine N-methyltransferase SUV39H1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6839
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.