Learn More
Invitrogen™ SUV39H1 Recombinant Superclonal™ Antibody (3HCLC)
Rabbit Recombinant Superclonal Antibody
Supplier: Invitrogen™ 711473
Description
This antibody is predicted to react with Monkey, Bovine, Sheep Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
This gene encodes an evolutionarily-conserved protein containing an N-terminal chromodomain and a C-terminal SET domain. The encoded protein is a histone methyltransferase that trimethylates lysine 9 of histone H3, which results in transcriptional gene silencing. Loss of function of this gene disrupts heterochromatin formation and may cause chromosome instability. Alternative splicing results in multiple transcript variants.
Specifications
SUV39H1 | |
Recombinant Superclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O43463 | |
SUV39H1 | |
Protein corresponding to human SUV39H1 (aa65-214). | |
100 μg | |
Primary | |
Human, Cynomolgus Monkey | |
Antibody | |
IgG |
Western Blot | |
3HCLC | |
Unconjugated | |
SUV39H1 | |
AI852103; AL022883; DXHXS7466e; H3-K9-HMTase 1; Histone H3-K9 methyltransferase 1; histone-lysine N-methyltransferase SUV39H1; histone-lysine N-methyltransferase, H3 lysine-9 specific 1; KMT1A; Lysine N-methyltransferase 1A; MG44; mIS6; Position-effect variegation 3-9 homolog; RGD1565028; Su(var)3-9 homolog 1; suppressor of variegation 3-9 homolog 1; suppressor of variegation 3-9 homolog 1 (Drosophila); SUV39H; Suv39h histone methyltransferase; SUV39H1 | |
Rabbit | |
Protein A | |
RUO | |
102134179, 6839 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.