Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUV39H2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102008
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102008 Supplier Invitrogen™ Supplier No. PA5102008
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SUV39H2.

The murine gene Suv39h2 encodes an H3 histone methyltransferase (HMTase) 59% identical in sequence to mouse Suv39h1. During embryogenesis, both proteins overlap in tissue expression, yet Suv39h2 transcripts are restricted to the testes in adult animals. Immunolocalization of the Suv39h2 protein during spermatogenesis indicates enrichment at the heterochromatin from the leptotene to the round spermatid stage. Moreover, Suv39h2 specifically accumulates with chromatin of the sex chromosomes, which undergo transcriptional silencing during the first meiotic prophase. Suv39h2 HMTase may also organize meiotic heterochromatin with the potential for epigenetic imprint to the male germline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUV39H2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SUV39H2
Gene Accession No. Q9EQQ0, Q9H5I1
Gene Alias 4930507K23Rik; AA536750; D030054H19Rik; D2Ertd544e; FLJ23414; H3 K9 HMTase 2; H3-K9-HMTase 2; Histone H3-K9 methyltransferase 2; histone-lysine N-methyltransferase SUV39H2; KMT1; KMT1B; lysine N-methyltransferase 1B; RP11-2K17.2; Su; Su(var)3-9 homolog 2; suppressor of variegation 3-9 homolog 2; suppressor of variegation 3-9 homolog 2 (Drosophila); Suv39h histone methyltransferase; Suv39h2
Gene Symbols SUV39H2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SUV39H2(Accession Q9H5I1), corresponding to amino acid residues K117-S167.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64707, 79723
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.