Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUV420h1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197313AF750
Description
SUV420h1 Polyclonal specifically detects SUV420h1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SUV420h1 | |
Polyclonal | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
C630029K18Rik, CGI85, CGI-85, EC 2.1.1.43, histone-lysine N-methyltransferase SUV420H1, KMT5B, KMT5B Lysine N-methyltransferase 5B, MGC118906, MGC118909, MGC21161, MGC703, Su(var)4-20 homolog 1, Suppressor of variegation 4-20 homolog 1, suppressor of variegation 4-20 homolog 1 (Drosophila), suv4-20h1 | |
Rabbit | |
Affinity Purified | |
Cancer, Epigenetics, Transcription Factors and Regulators | |
51111.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
KMT5B | |
A synthetic peptide made to an internal portion of the human SUV420h1 protein (between residues 450-500) [UniProt Q4FZB7] | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction