Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUV420h1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197313H
Description
SUV420h1 Polyclonal specifically detects SUV420h1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative).Specifications
SUV420h1 | |
Polyclonal | |
Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative) | |
Q4FZB7 | |
KMT5B | |
A synthetic peptide made to an internal portion of the human SUV420h1 protein (between residues 450-500) [UniProt Q4FZB7] | |
0.1 mL | |
Cancer, Epigenetics, Transcription Factors and Regulators | |
51111 | |
Store at 4C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
HRP | |
PBS with No Preservative | |
C630029K18Rik, CGI85, CGI-85, EC 2.1.1.43, histone-lysine N-methyltransferase SUV420H1, KMT5B, KMT5B Lysine N-methyltransferase 5B, MGC118906, MGC118909, MGC21161, MGC703, Su(var)4-20 homolog 1, Suppressor of variegation 4-20 homolog 1, suppressor of variegation 4-20 homolog 1 (Drosophila), suv4-20h1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction