Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUV420h1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299518PCP
Description
SUV420h1 Polyclonal antibody specifically detects SUV420h1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SUV420h1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C630029K18Rik, CGI85, CGI-85, EC 2.1.1.43, histone-lysine N-methyltransferase SUV420H1, KMT5B, KMT5B Lysine N-methyltransferase 5B, MGC118906, MGC118909, MGC21161, MGC703, Su(var)4-20 homolog 1, Suppressor of variegation 4-20 homolog 1, suppressor of variegation 4-20 homolog 1 (Drosophila), suv4-20h1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SUV420h1. | |
0.1 mL | |
Cancer, Epigenetics, Transcription Factors and Regulators | |
51111 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction