Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUV420H1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566611 Supplier Invitrogen™ Supplier No. PA566611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DQGEHSGTVGV PVSYTDCAPS PVGCSVVTSD SFKTKDSFRT AKSKKKRRIT RYDAQLILEN NSGIPKLTLR Highest antigen sequence identity to the following orthologs - mouse 86%, rat 87%.

Histone methyltransferase that specifically trimethylates 'Lys-20' of histone H4. H4 'Lys-20' trimethylation represents a specific tag for epigenetic transcriptional repression. Mainly functions in pericentric heterochromatin regions, thereby playing a central role in the establishment of constitutive heterochromatin in these regions. KMT5B is targeted to histone H3 via its interaction with RB1 family proteins (RB1, RBL1 and RBL2). Plays a role in myogenesis by regulating the expression of target genes, such as EID3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUV420H1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KMT5B
Gene Accession No. Q4FZB7
Gene Alias [histone H4]-lysine20 N-methyltransferase KMT5B; [histone H4]-N-methyl-L-lysine20 N-methyltransferase KMT5B; AA117471; C630029K18Rik; CGI85; CGI-85; histone-lysine N-methyltransferase KMT5B; histone-lysine N-methyltransferase SUV420H1; Kmt5b; lysine (K)-specific methyltransferase 5B; lysine methyltransferase 5B; lysine N-methyltransferase 5B; Lysine-specific methyltransferase 5B; MGC118906; MGC118909; MGC21161; MGC703; si:dkey-12e7.2; Su(var)4-20 homolog 1; suppressor of variegation 4-20 homolog 1; suppressor of variegation 4-20 homolog 1 (Drosophila); suppressor of variegation 4-20 protein-like 1; SUV420; SUV420H1; suv4-20h1; Unknown (protein for MGC:137299); wu:fb97g06; wu:fi57g03; zgc:103527
Gene Symbols KMT5B
Host Species Rabbit
Immunogen Recombinant Human SUV420H1. Recombinant protein control fragment (Product #RP-107271).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51111
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.