Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUV420H2 Antibody (RAB-C409), Janelia Fluor™ 646 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320151JF646
Description
SUV420H2 Monoclonal antibody specifically detects SUV420H2 in Human samples. It is validated for ELISASpecifications
SUV420H2 | |
Monoclonal | |
Janelia Fluor 646 | |
EC 2.1.1.43, FLJ98627, histone-lysine N-methyltransferase SUV420H2, KMT5Csuv4-20h2, Lysine N-methyltransferase 5C, MGC2705, Su(var)4-20 homolog 2, Suppressor of variegation 4-20 homolog 2, suppressor of variegation 4-20 homolog 2 (Drosophila), Suv4-20h2 | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing SUV420H2 protein under non-denaturing conditions. | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
RAB-C409 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
84787 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction