Learn More
Invitrogen™ SUV420H2 Chimeric Recombinant Rabbit Monoclonal Antibody (RAB-C409)

Description
Specificity: This antibody recognizes KMT5C (Histone-lysine N-methyltransferase KMT5C). It binds to a folded domain, amino acids 2-248. KMT5C is a histone methyltransferase that regulates transcription and maintenance of genome integrity.
Specifications
Specifications
| Antigen | SUV420H2 Chimeric |
| Applications | ChIP Assay, ELISA, Flow Cytometry, Immunoprecipitation, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | RAB-C409 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | KMT5C |
| Gene Accession No. | Q86Y97 |
| Gene Alias | BC024816; Histone-lysine N-methyltransferase KMT5C; histone-lysine N-methyltransferase SUV420H2; KMT5C; lysine (K)-specific methyltransferase 5C; lysine methyltransferase 5C; lysine N-methyltransferase 5C; Lysine-specific methyltransferase 5C; PP7130; Su(var)4-20 homolog 2; suppressor of variegation 4-20 homolog 2; suppressor of variegation 4-20 homolog 2 (Drosophila); SUV420H2; suv4-20h2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.