Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUV420H2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145093 Supplier Invitrogen™ Supplier No. PA5145093
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: THKMNVSPVP PLRRQQHLRS ALETFLRQRD LEAAYRALTL GGWTARYFQS R Antigen sequence identity: Mouse 88%; Rat 86%.

SUV420H2 is a histone methyltransferase that specifically trimethylates 'Lys-20' of histone H4. H4 'Lys-20' trimethylation represents a specific tag for epigenetic transcriptional repression. The protein mainly functions in pericentric heterochromatin regions, thereby playing a central role in the establishment of constitutive heterochromatin in these regions. SUV420H1 is targeted to histone H3 via its interaction with RB1 family proteins (RB1, RBL1 and RBL2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUV420H2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KMT5C
Gene Accession No. Q86Y97
Gene Alias BC024816; Histone-lysine N-methyltransferase KMT5C; histone-lysine N-methyltransferase SUV420H2; KMT5C; lysine (K)-specific methyltransferase 5C; lysine methyltransferase 5C; lysine N-methyltransferase 5C; Lysine-specific methyltransferase 5C; PP7130; Su(var)4-20 homolog 2; suppressor of variegation 4-20 homolog 2; suppressor of variegation 4-20 homolog 2 (Drosophila); SUV420H2; suv4-20h2
Gene Symbols KMT5C
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 31-81. Recombinant protein control fragment (Product #RP-110078).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84787
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.