Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUZ12 Antibody (2B9G12) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP261892

 View more versions of this product

Catalog No. NBP2-61892


Only null left
Add to Cart

Description

Description

SUZ12 Monoclonal specifically detects SUZ12 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

SUZ12
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
ChET 9 protein, CHET9polycomb protein SUZ12, Chromatin precipitated E2F target 9 protein, JJAZ1Joined to JAZF1 protein, KIAA0160Suppressor of zeste 12 protein homolog, suppressor of zeste 12 homolog (Drosophila)
Mouse
83 kDa
0.1 mL
23512
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
2B9G12
Unconjugated
PBS with 0.05% Sodium Azide
SUZ12
Purified recombinant fragment of human SUZ12 (AA: 1-139) expressed in E. Coli.
Protein G purified
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.