Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUZ12 Mouse anti-Human, Clone:2B9G12, Abnova™

Catalog No. 89333565 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-565 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-565 Supplier Abnova Corporation Supplier No. MAB17308
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human SUZ12.

This zinc finger gene has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region. [provided by RefSeq]

Specifications

Antigen SUZ12
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2B9G12
Conjugate Unconjugated
Description suppressor of zeste 12 homolog (Drosophila)
Dilution ELISA (1:10000)
Flow Cytometry (1:200-400)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene SUZ12
Gene Alias CHET9, JJAZ1, KIAA0160
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-139 of of human SUZ12 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 23512
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.