Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SUZ12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584345
Description
Immunogen sequence: NKPGSVKPTQ TIAVKESLTT DLQTRKEKDT PNENRQKLRI FYQFLYNNNT RQQTEARDDL HCPWCTLNCR KLYSLLKHLK LCHSRFIFNY VYHPKGARID VSINECYDGS YAGNPQDIHR QPGFAFSRNG PVKRTPIT.
SUZ12 (polycomb protein SUZ12) is a zinc finger gene and has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. SUZ12 is also a component of the PRC2/EED-EZH2 complex which methylates Lys-9 (H3K9me) and Lys-27 (H3K27me) of histone H3 leading to transcriptional repression of the affected target gene. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region.
Specifications
SUZ12 | |
Polyclonal | |
Unconjugated | |
SUZ12 | |
2610028O16Rik; AI195385; AU016842; AW536442; chET 9 protein; CHET9; chromatin precipitated E2F target 9 protein; D11Ertd530e; JJAZ1; Joined to JAZF1 protein; Kiaa0160; mKIAA0160; Polycomb protein Suz12; suppressor of zeste 12 homolog; suppressor of zeste 12 homolog (Drosophila); suppressor of zeste 12 protein homolog; Suz12; SUZ12 polycomb repressive complex 2 subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23512 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15022 | |
SUZ12 | |
Recombinant protein corresponding to Human SUZ12. Recombinant protein control fragment (Product #RP-102304). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction