Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Suz12 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992692
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
50 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2692 50 μL
50-199-2691 10 μL
50-199-2690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-199-2692 Supplier Active Motif Supplier No. 39057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SUZ12 (polycomb protein SUZ12) is a zinc finger gene and has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. SUZ12 is also a component of the PRC2/EED-EZH2 complex which methylates Lys-9 (H3K9me) and Lys-27 (H3K27me) of histone H3 leading to transcriptional repression of the affected target gene. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region.

Specifications

Antigen Suz12
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene SUZ12
Gene Accession No. Q15022, Q80U70
Gene Alias 2610028O16Rik; AI195385; AU016842; AW536442; ChET 9 protein; CHET9; Chromatin precipitated E2F target 9 protein; D11Ertd530e; JJAZ1; Joined to JAZF1 protein; KIAA0160; mKIAA0160; Polycomb protein Suz12; suppressor of zeste 12 homolog (Drosophila); Suppressor of zeste 12 protein homolog; SUZ12; SUZ12 polycomb repressive complex 2 subunit
Gene Symbols SUZ12
Host Species Rabbit
Immunogen This Suz12 antibody was raised against a peptide corresponding to the C-terminus of human Suz12.
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23512, 52615
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.