Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUZ12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 203661AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-2851


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

This zinc finger gene has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region.
Specifications

Specifications

SUZ12
Polyclonal
Unconjugated
SUZ12
ChET 9 protein, CHET9, JJAZ1, Joined to JAZF1 protein, KIAA0160, Polycomb protein SUZ12, SUZ12
Rabbit
Antigen Affinity Chromatography
RUO
23512, 52615, 688041
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15022, Q80U70
Suz12
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.