Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUZ12 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | SUZ12 |
---|---|
Dilution | Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB126787
|
Novus Biologicals
NBP310944100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
SUZ12 Polyclonal specifically detects SUZ12 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry.Specifications
SUZ12 | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
23512 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
ChET 9 protein, CHET9polycomb protein SUZ12, Chromatin precipitated E2F target 9 protein, JJAZ1Joined to JAZF1 protein, KIAA0160Suppressor of zeste 12 protein homolog, suppressor of zeste 12 homolog (Drosophila) | |
The immunogen is a synthetic peptide directed towards the middle region of human SUZ12 (NP_056170). Peptide sequence TGETNDKSTAPIAKPLATRNSESLHQENKPGSVKPTQTIAVKESLTTDLQ | |
Affinity purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title