Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SV2B Monoclonal Antibody (2C6)
GREENER_CHOICE

Catalog No. PIMA524430
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524430 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524430 Supplier Invitrogen™ Supplier No. MA524430
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: YFQDEEYKSK MKVFFGEHVY GATINFTMEN QIHQHGKLVN DKFTRMYFKH VLFEDTFFDE.

Probably plays a role in the control of regulated secretion in neural and endocrine cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SV2B
Applications ELISA
Classification Monoclonal
Clone 2C6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Sv2b
Gene Accession No. Q7L1I2
Gene Alias A830038F04Rik; AI850768; HsT19680; Kiaa0735; mKIAA0735; SV2B; Sv2bb; synaptic vesicle glycoprotein 2 b; synaptic vesicle glycoprotein 2b; Synaptic vesicle protein 2B; synaptic vesicle protein 2B homolog
Gene Symbols Sv2b
Host Species Mouse
Immunogen SV2B (NP_055663, 417 a.a. ~ 476 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9899
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.