Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SV2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561194
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561194 Supplier Invitrogen™ Supplier No. PA561194
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EYQGIPHPDD VKAKQAKMAP SRMDSLRGQT DLMAERLEDE EQLAHQYETI MDECGHGR Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 45%.

Probably plays a role in the control of regulated secretion in neural and endocrine cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SV2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sv2b
Gene Accession No. Q7L1I2
Gene Alias A830038F04Rik; AI850768; HsT19680; Kiaa0735; mKIAA0735; SV2B; Sv2bb; synaptic vesicle glycoprotein 2 b; synaptic vesicle glycoprotein 2b; Synaptic vesicle protein 2B; synaptic vesicle protein 2B homolog
Gene Symbols Sv2b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SV2B. Recombinant protein control fragment (Product #RP-98176).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9899
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.