Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SV2B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981304
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1304 Supplier Bioss Supplier No. BS11365R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Probably plays a role in the control of regulated secretion in neural and endocrine cells.

Specifications

Antigen SV2B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Sv2b
Gene Accession No. Q7L1I2
Gene Alias A830038F04Rik; AI850768; BOS_20015; HsT19680; KIAA0735; LOC100342160; mKIAA0735; SV2B; Sv2bb; synaptic vesicle glycoprotein 2 b; Synaptic vesicle glycoprotein 2B; Synaptic vesicle protein 2B; synaptic vesicle protein 2B homolog
Gene Symbols Sv2b
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SV2B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9899
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.