Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SV2C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559290
Description
Immunogen sequence: GEYQGIPSMN QAKDSIVSVG QPKGDEYKDR RELESERRAD EEELAQQYE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles.
Specifications
SV2C | |
Polyclonal | |
Unconjugated | |
SV2C | |
4930527L09Rik; AI841971; KIAA1054; SV2C; Synaptic vesicle glycoprotein 2C; synaptic vesicle protein 2C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q496J9 | |
SV2C | |
Recombinant protein corresponding to Human SV2C. Recombinant protein control fragment (Product #RP-96839). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction