Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SV2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572189
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572189 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572189 Supplier Invitrogen™ Supplier No. PA572189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SV2C Polyclonal Antibody for Western Blot

Plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SV2C
Applications Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SV2C
Gene Accession No. Q496J9, Q9Z2I6
Gene Alias 4930527L09Rik; AI841971; KIAA1054; SV2C; Synaptic vesicle glycoprotein 2C; synaptic vesicle protein 2C
Gene Symbols SV2C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 547-580 amino acids from the C-terminal region of human SV2C.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22987, 29643
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.